Learn More
Fisherbrand™ Borosilicate Glass Burette Tips
Add to the end of a burette to perform volumetric titration applications. Fisherbrand™ Borosilicate Glass Burette Tips fit single bore burettes.
Brand: Fisherbrand™
Additional Details : Weight : 0.04500kg
Description
- Compatible with Fisherbrand™ single bore burettes
- Borosilicate glass
- For use with titration applications
Specifications
Burette | |
Borosilicate Glass | |
Buret Clamp Tip |