Learn More
JG Finneran Associates™ Borosilicate Glass Shell Vials
Use these vials for a variety of applications. JG Finneran™ Borosilicate Glass Shell Vials feature thicker walls for safer sample handling.
Brand:  JG Finneran Associates™ 4100-1232
 View more versions of this product
Product Code. 10422751
  • 121.00 SEK / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Glass shell vial
  • Available in clear or amber Type I borosilicate glass for easy disposal
  • Limited volume shell vials have a precisely designed conical bottom which permits maximum sample evacuation
Specifications
Specifications
Clear
2 mL
100 Pk.
Borosilicate Glass
100
12 mm