Learn More
MP Biomedicals Briggs Chick Salt Mixture No. 5
Salt mixture for chicks. Contains different variety of salts.
Brand: MP Biomedicals 0290283610
This item is not returnable.
View return policy
Description
Salt mixture containing calcium carbonate, calcium phosphate, etc.Specifications
Not Autoclavable | |
Single-use | |
Non-sterile | |
1 ft. |
Animal Diet | |
10 kg | |
Room Temperature | |
0.3048 m |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.