Learn More
C14orf106 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26264-1-AP-20UL
This item is not returnable.
View return policy
Description
MIS18BP1 is required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis.Specifications
C14orf106 | |
Polyclonal | |
Unconjugated | |
MIS18BP1 | |
C14orf106, HSA242977, HsKNL 2, KIAA1903, KNL2, M18BP1, Mis18 binding protein 1, MIS18BP1, P243 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55320 | |
-20°C | |
Liquid |
Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6P0N0 | |
MIS18BP1 | |
C14orf106 Fusion Protein Ag23483 | |
20 μL | |
Primary | |
Human | |
Antibody | |