Läs mer
Swann-Morton™ Carbon Steel Individual Peel Pack, Non-Sterile Scalpel Blades, No. 11
Kvantitet:
100/kartong
Förpackningsstorlek:
Paket med 100
Beskrivning
Elongated triangular blade sharpened along the hypotenuse edge with a strong pointed tip making it ideal for stab incisions.
- Elongated triangular blade.
- Sharpened along the hypotenuse edge.
- Strong, pointed tip, making it ideal for stab incisions needed when lancing an abscess or inserting a chest drain.
- It is held like a pencil and often upside down by the surgeon to prevent it inadvertently being inserted too deeply.
- To avoid this scenario, the scalpel should not be held towards the back end whilst the heel of the hand should be placed on the skin thus stabilizing the hand to help prevent an unwanted deep incision.
- The No.11 is also used for other small incisions such as when inserting a laparoscopic port, vein and artery central lines, opening the aorta and removing calcifications in the aortic or mitral valves, anatomical dissections, and surgical treatment of disorders of the foot, ankle, and lower extremities in podiatry.
- Fits handles 3, 3L, 3 Graduated, 5B, 7, 9 and B3.
Specifikationer
Specifikationer
| Bladmaterial | Kolstål |
| Sterilitet | Icke-steril |
| Kvantitet | 100/kartong |
| Förpackningstyp | Individuellt förpackad |