missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Casein, Vitamin Tested, 30 Mesh

Used as cheese, food additive and binder for safety matches.

Brand:  MP Biomedicals™ 0290452001

 View more versions of this product

Product Code. 11445722

  • 723.00 SEK / 1lb

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Phosphoprotein
  • Extracted from bovine milk
  • Used as food source
  • CAS: 9000-71-9
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Casein
Powder
1 lb.
Yellow
Used as cheese, food additive and binder for safety matches
Store at room temperature (15°C to 30°C)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers