missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA531822
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Recommended positive controls: NT2D1, IMR32, U87-MG. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May play a role in tumorigenesis.
Specifica
| CCDC7 | |
| Polyclonal | |
| Unconjugated | |
| CCDC7 | |
| BIOT2; BioT2-A; BioT2-B; BioT2-C; C10orf68; CCDC7; coiled-coil domain containing 7; Coiled-coil domain-containing protein 7; coiled-coil domain-containing protein 7; uncharacterized protein C10orf68; Protein BIOT2; RP11-479G22.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79741 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96M83 | |
| CCDC7 | |
| Recombinant protein encompassing a sequence within the center region of human CCDC7. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto