Läs mer
Eurofins DiscoverX Cdk7/Cyclin H/MAT1 (CAK complex) Protein, Active
Beskrivning
Cyclin-dependent Kinase 7 (cdk7) is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This protein forms a trimeric complex with cyclin H and MAT1, which functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This protein is thought to serve as a direct link between the regulation of transcription and the cell cycle.
GenBank X79193 / GenBank U11791 / GenBank X87843
Specifikationer
Specifikationer
| Formulering | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molekylvikt (g/mol) | 39.9 kDa (CDK7), 37.8 kDa (Cyclin H) and 62.8 kDa (MAT1) |
| Kvantitet | 250 μg |
| Art | Human |
| Rekombinant | Recombinant |
| Protein Tag | C-terminal 6His / untagged / N-terminal GST |
| Uttryckssystem | Recombinant enzyme expressed in Sf21 insect cells |
| Proteinform | Full-length |
| Renhet eller kvalitetsklass | Combined ≥54% |
| Protein | CDK7/Cyclin E/MAT1, active |
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.