missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Cell Culture Dishes, Non-Treated
For cell based assays and microscopic analysis. Eppendorf™ Cell Culture Dishes, Non-Treated helps in direct access to the cells.
Specifications
Color | Transparent |
---|---|
Material | Polystyrene |
Sterility | Sterile |
Surface Treatment | Non-treated |
Type | Cell Culture Dish |
Product Code | Brand | dimDiameterMetric | Volume (Metric) Working | Culture Area | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | dimDiameterMetric | Volume (Metric) Working | Culture Area | Price | Quantity & Availability | |||
15690507
|
Eppendorf™
0030700015 |
2 to 3 mL | 9.5 sq. cm |
N/A
|
N/A | ||||
15610517
|
Eppendorf™
0030701011 |
3 to 5 mL | 21.9 sq. cm |
N/A
|
N/A | ||||
15630517
|
Eppendorf™
0030702018 |
8 to 10 mL | 56.8 cm2 |
N/A
|
N/A | ||||
Description
- Safety in handling during transportation and while working due to corrugated handling ring
- SplashProtect ring inside of the dish lid traps liquid and prevents spills during transportation or incubation
- Pronounced rim on dish lid and exact fitting of lid and bottom ensure sturdy, secure stacking
- Easy differentiation of lid and dish bottom prevents unintended removal of the lid
- Packaging combines tool-free opening, resealable top and side for tape-free closing, and an easy solution to shrink the package for space-conscious storage
- Available in three formats: 35, 60 and 100mm
Specifications
Transparent | |
Sterile | |
Cell Culture Dish |
Polystyrene | |
Non-treated |