Learn More
TPP™ Cell Scrapers
TPP™ cell scraper require slight pressure on handle using forefinger, thus pushing handle downward towards floor of container.
1870.90 SEK - 3646.40 SEK
Product Code | Brand | For Use With (Equipment) | Width (Metric) Blade | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Width (Metric) Blade | Price | Quantity & Availability | ||||
11391714
|
TPP™
99003 |
Tissue culture flask T-75 / T-150, tissue culture dish 100/150 | 20 mm |
3646.40 SEK
Pack of 150 |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
11301724
|
TPP™
99004 |
Tissue culture flask T-300 | 25 mm |
1870.90 SEK
Pack of 100 |
Estimated Shipment: 17-10-2024
Log in to see stock. |
||||
Description
TPP™ cell scraper require slight pressure on handle using forefinger, thus pushing handle downward towards floor of container.
- Free rotating blade
- Total access to all corner
- Different blade widths
- Small raised knobs on handle
- Lint-free blister packaging
- User friendly TPP dispenser case
Specifications
Cell Scraper | |
Polyethylene |