missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ ChromatoPur™ Bovine Thrombin, Low Specific Activity, Diagnostic Grade
Suitable as a diagnostic and research reagent in clotting assays
Brand: MP Biomedicals™ 0219990725
This item is not returnable.
View return policy
Description
Chromatographically purified with minimal loss of native clotting activity and minimized degradation to the gamma form. Enzyme activity is measured by clotting assay. 90 to 300U/mg protein.
Physical Characteristics
- Appearance: White to off-white powder
- pH: 7.0 to 8.0
- Protein: ≥50%
- Specific activity: 90 to 300U/mg protein
- Activity: Report μ/mg solid
- Total Activity: U/vial
- Exipients: Reported
Specifications
Cell Culture, Microbial Culture | |
7 to 8 | |
Bovine Thrombin | |
Solid |
Bovine Thrombin | |
25 kU | |
-10°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Bovine Thrombin, Low Specific Activity, Diagnostic Use
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Thrombin
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction