Learn More
cis-2-Pentene, Thermo Scientific Chemicals
720.68 SEK - 3396.29 SEK
Chemical Identifiers
CAS | 627-20-3 |
---|---|
Molecular Formula | C5H10 |
Molecular Weight (g/mol) | 70.14 |
MDL Number | MFCD00063978 |
InChI Key | QMMOXUPEWRXHJS-HYXAFXHYSA-N |
Synonym | cis-2-pentene, z-2-pentene, 2-pentene, z, cis-pentene, z-pent-2-ene, unii-54ur3xz4fc, cis-pentene-2, 54ur3xz4fc, 2-pentene, 2-pentene, 2z |
PubChem CID | 5326160 |
IUPAC Name | (Z)-pent-2-ene |
SMILES | CC\C=C/C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10320684
|
Thermo Scientific Acros
383590010 |
1 g | Glass bottle |
720.68 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10299573
|
Thermo Scientific Acros
383590050 |
5 g | Glass bottle |
2165.58 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10116823
|
Thermo Scientific Acros
383590100 |
10 g | Glass bottle |
3396.29 SEK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
627-20-3 | |
70.14 | |
QMMOXUPEWRXHJS-HYXAFXHYSA-N | |
5326160 | |
CC\C=C/C |
C5H10 | |
MFCD00063978 | |
cis-2-pentene, z-2-pentene, 2-pentene, z, cis-pentene, z-pent-2-ene, unii-54ur3xz4fc, cis-pentene-2, 54ur3xz4fc, 2-pentene, 2-pentene, 2z | |
(Z)-pent-2-ene |
Specifications
627-20-3 | |
Colorless | |
37.0°C | |
Authentic | |
CH3CHCHCH2CH3 | |
MFCD00063978 | |
0.65 | |
Solubility in water: insoluble. | |
CC\C=C/C | |
70.14 | |
70.13 | |
Liquid |
-151.0°C | |
0.6500g/mL | |
−27°C | |
C5H10 | |
1.3810 to 1.3830 (20°C, 589nm) | |
15, 7235 | |
cis-2-pentene, z-2-pentene, 2-pentene, z, cis-pentene, z-pent-2-ene, unii-54ur3xz4fc, cis-pentene-2, 54ur3xz4fc, 2-pentene, 2-pentene, 2z | |
QMMOXUPEWRXHJS-HYXAFXHYSA-N | |
(Z)-pent-2-ene | |
5326160 | |
≥95.0% | |
cis-2-Pentene |
Safety and Handling
GHS H Statement:
May be fatal if swallowed and enters airways.
Harmful if swallowed.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 210-988-7
- cis-2-Pentene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Aspiration toxicity Category 1
- Flammable liquid Category 2
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P331-Do NOT induce vomiting.
RUO – Research Use Only