Läs mer
Electrothermal™ CMUT Series Multi-Volume Controlled Mantle
Kapacitet (metrisk):
250 till 1000 ml
Förpackningsstorlek:
Varje
Beskrivning
Accommodates round bottom flasks between 250 to 1000mL capacity. Electrothermal™ CMUT Series Multi-Volume Contiolled Mantle is used in wet chemistry to heat liquids in round bottom flasks.
- Metal case is designed to remain “Cool-to-the-touch” when in operation.
- For added safety, it has a grounded stainless steel earth screen covering the element and is double fused.
- Multiple volume round bottom flask capacity between 250 to 1000mL
- Element temperatures up to 450°C
- Built-in energy regulator
- Indicator lamps for power and heater operation
- Replaceable insulated heater cartridge
- Rugged and easy to clean powder coated aluminum outer casing
- Non-skid feet and support rod bracket (Support rods not included)
- Flexible coiled heating elements absorb shock, minimising the risk of flask breakage
- Heating element is attached to a rigid thermal insulating cartridge to ensure maximum heat transfer and support
Specifikationer
Specifikationer
| Kapacitet (metrisk) | 250 till 1000 ml |
| Mått (L x B x H) | 220 x 220 x 170mm |
| Spänning | 230 V |
| Wattal | 300W |
| Vikt (metrisk) Frakt | 1.5kg |
| Temperatur (metrisk) | 450°C |
| Frekvens | 50/60 Hz |