Läs mer
Contec™ Anticon™ Gold Sorb Cleanroom Wipes
Combine the performance of microfiber yarn with that of a polyester yarn for superior wiping and sorptive characteristics. Contec™ Anticon™ Gold Sorb Cleanroom Wipes feature proprietary sealed edges that allow the use of the entire wipe surface without the worry of harming delicate surfaces.
Brand:  Contec™ 492238-957
Produktkod. 13141056
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
  • Combine the performance of microfiber and polyester for superior wiping and sorptive characteristics
  • No abrasive borders, which makes wipes particulary suitable for cleaning sensitive components such as optics
  • Sealed edges reduce particle and fiber generation
  • Unique microfiber provides superior residue and particle removal even when dry
  • High sorptive capacity and fast adsorption rate for superior spill control
  • Cleanroom laundered
  • Tested in accordance with the IEST Recommend Practices for the Evaluation of Wiping Materials for Critical Environments
  • 12 x 12 in. wipes available sterile 