Learn More
Conventional Closure Polyethylene Terephthalate Blood Tube
Brand: BD Vacutainer 367526
276.30 SEK valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Additional Details : Weight : 0.86850kg
Specifications
Blood Collection Tube | |
PET | |
10 mL | |
Paper |
Lithium Heparin | |
Traditional Cap | |
Green | |
16 mm |