Learn More
Corning™ BioCoat™ Matrigel™ Invasion Chamber with Corning™ Matrigel Matrix
GREENER_CHOICE
Provide rapid and reproducible quantification of cell invasion in vitro. Corning™ BioCoat™ Matrigel™ Invasion Chamber with Corning Matrigel Matrix™ cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and differentiation.
Brand:  Corning™ 354480
6691.87 SEK valid until 2024-11-29
Use promo code "21683" to get your promotional price.
 View more versions of this product
Product Code. 11553570
  • 7443.67 SEK / Pack of 24
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Description
Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
Coatings include Corning Matrigel™ matrix, Fibronectin, Collagen, and Laminin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24
PET
Studies in which minimal interference by growth factors is desirable
12/Pk.
Matrigel™ Matrix
0.3 cm2
8 μm
2
24/Cs.