Läs mer
Cole-Parmer™ CPC Miniature Quick-Disconnect Valved Male Hose Barb
Produktkod. 15466523
Hose Barb ID:
1/8 tum
Förpackningsstorlek:
Paket med 10
Produktkod. 15466523
Brand: Cole-Parmer™ 0636384
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Quick Disconnect couplings feature a thumb latch for easy assembly.
  • CPC Miniature Quick-Disconnect Valved Male Hose Barb
  • Standard flow quick-disconnect couplings require a body and an insert of the same flow size
  • Plug insert into body to connect fitting
  • Fitting halves lock into place immediately
  • To disconnect fitting halves, simply press the thumb latch on the coupling body
Specifikationer
Beskrivning Quick-Disconnect x Barb Connection
Material Polycarbonate
Total längd (metrisk) 2.286 cm
Slangstorlek 1/8 in.
Utloppsport 3/32 in.
Hose Barb ID 1/8 tum
Produkttyp Valved Male Hose Barb