Läs mer
Cole-Parmer™ CPC™ Sanitetskontakt, PE-hylsa, encykel
Klicka för att se tillgängliga alternativ
Diameter (engelska) Inre:
1/2 tum
3/8 tum
Förpackningsstorlek:
Varje
Beskrivning
- Disposable single cycle sleeve.
- Save time and money on cleaning.
- Eliminates dead legs where bacteria can grow.
- Connects to most bio-bag connections.
- Steam-through connections will transform your steam-in place (SIP) process by making the connections cleaner, faster and easier.
- Innovative product design allows for a one-step process for sterilization and media transfer while eliminating dead spots, reducing the number of connections and eliminating the need for a laminar flow hood.
- The terminations on the fittings are set to industry standards for bio-reactors and popular sizes of flexible tubing, speeding the adaptation to existing systems and processes.
- Tear-away sleeve protects the fitting from actuation prior to complete sterilization.
- The polysulfone body and silicone membrane materials meet requirements of USP Class VI, and physicochemical tests.
Specifikationer
Specifikationer
| Beskrivning | Sanitary x Barb |
| Diameter (engelska) Inre | 3/8 tum |
| Material | Polysulfone, PE Sleeve |
| Slangstorlek | 3/4 in. |
| Slang Barb | 3/4 in. |
| Utloppsport | 3/4 in. |
| Steriliseringsmetod | Gamma or steam-in-place (SIP) |
| Produkttyp | Sanitary Connector, Single-Cycle |