missing translation for 'onlineSavingsMsg'
Learn More

Creatine Phosphate, Disodium Salt Tetrahydrate, MP Biomedicals™

Product Code. 11417580
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Creatine phosphate is substrate for CPK determination.

TRUSTED_SUSTAINABILITY

Specifications

CAS 922-32-7
pH 7 to 9 (1% aq. soln.)
Molecular Formula C4H8N3Na2O5P
MDL Number MFCD00150192
Quantity 5 g
Synonym creatine phosphate sodium salt
Solubility Information Soluble in water. (150mg/mL - clear, colorless solution)
InChI Key RNTXMYSPASRLFT-UHFFFAOYSA-L
SMILES [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
IUPAC Name disodium ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
Molecular Weight (g/mol) 255.08
PubChem CID 131887655
Formula Weight 327.2
Percent Purity ≥98%
Physical Form Powder
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'