Learn More
TPP™ Cryo Tubes
TPP™ Cryo Tubes are used to store samples in freezers or similar low-temperature equipment.
Brand:  TPP™ 89020
Product Code. 11301734
  • 3720.00 SEK / Pack of 800
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
TPP™ Cryo Tubes are used to store samples in freezers or similar low-temperature equipment.
  • External thread
  • Colour-coding inserts in a range of colours
  • Abstain from using silicone gasket, enabling secure and tight closure of tube
  • Puncture and tear resistant re-closable bags with zipper and laser perforation
  • Self-standing, star foot
  • Graduated with large writing surface
Specifications
Specifications
Cryogenic Storage Tube
1.5 mL
800
Dimensions: 12 x 48 mm
Polypropylene