Learn More
Epredia™ Cryotome™ Cryostat Accessories, Camel Hair Brush
Enhance the operation of Cryotome™ FSE/FE/SME/E Cryostats with Epredia™ Cryotome™ Cryostat Accessories.
Brand: Epredia™ 1910
384.56 SEK valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
Camel hair brush with wooden handle.Specifications
Camel Hair Brush | |
FSE/FE/SME/E Cryostats | |
Pk. of 12 | |
7.5 in. |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido