missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ CXCL17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122057
This item is not returnable.
View return policy
Description
CXCL17 is a chemokine that acts as chemoattractant for monocytes, macrophages and dendritic cells. In particular, CXCL17 is a mucosal chemokine that attracts dendritic cells and blood monocytes to the lungs. CXCL17 plays a role in angiogenesis and possibly in the development of tumors. Further, CXCL17 acts as an anti-inflammatory in the stomach and may play a role in the innate defense against infections. CXCL17 also activates the C-X-C chemokine receptor GPR35 to induce a rapid and transient rise in the level of intracellular calcium ions. 4-Cys CXCL17 seems to exhibit much higher chemoattractant potency on monocytes and macrophages than 6-Cys CXCL17. Diseases associated with CXCL17 dysfunction include cholera.
Specifications
| CXCL17 | |
| Polyclonal | |
| Unconjugated | |
| CXCL17 | |
| 13.6 kDa protein; 13.6-kDa protein; 4-Cys CXCL17; 6-Cys CXCL17; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 17; CXC; CXC motif chemokine; C-X-C motif chemokine 17; C-X-C motif chemokine ligand 17; CXCL; CXCL17; Dcip1; Dendritic cell and monocyte chemokine-like protein; DMC; UNQ473; UNQ473/PRO842; Vcc1; VCC-1; VEGF coregulated chemokine 1; VEGF co-regulated chemokine 1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 284340 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6UXB2 | |
| CXCL17 | |
| Leu24-Leu119 (Accession Q6UXB2), with N-terminal His and GST Tag. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction