missing translation for 'onlineSavingsMsg'
Learn More

α-D-(+)-Melibiose Monohydrate, MP Biomedicals™

Product Code. 11413490
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

α-D-(+)-Melibiose Monohydrate used to improve the preparation of benzoylated ethyl 1-thioglycosidesto and to study amorphous disaccharides.

  • Melibiose has been used in a study to improve the preparation of benzoylated ethyl 1-thioglycosides.
  • It has also been used in a study to investigate the effect of water plasticization on the molecular mobility and crystallization tendency of amorphous disaccharides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-99-9
Molecular Formula C12H22O11·H2O
Synonym 6-O-α-D-galactopyranosyl-D-glucose, α-D-melibiose hydrate

Specifications

Color White
Quantity 100 g
Optical Rotation +135 ± 10° (c=4, water)
Formula Weight 360.3
Percent Purity 99.9%
Physical Form Powder
Chemical Name or Material α-D-(+)-Melibiose monohydrate

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.