Läs mer
Digoxigenin Antibody (21H8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP2-31191
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Digoxigenin Monoclonal specifically detects Digoxigenin in All Species samples. It is validated for Western Blot, ELISA, Immunohistochemistry, In-situ Hybridization, Southern Blot.
Specifikationer
| Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
| Purified |
Korrigering av produktinnehåll
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering