Learn More
Nexcelom Bioscience Disposable Plastic Cell Counting Chambers
Disposable, standard slide thickness cell counting chamber slide. Nexcelom™ Disposable Plastic Cell Counting Chambers are designed with two counting chambers per slide.
Brand: Nexcelom Bioscience CHT4-SD100-002
Description
Disposable, standard slide thickness cell counting chamber slide. Nexcelom™ Disposable Plastic Cell Counting Chambers are designed with two counting chambers per slide.
- Standard slide thickness
- For cells ranging from 2 to 80μm in diameter
- Each slide has a protective film on both sides that must be removed before use
- Cost affective
- Disposable
- Convenient
- Two counting chambers per slide
- Box of 75 slides for 150 counts
Specifications
Cell Counting Chamber | |
Plastic | |
75 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.