Learn More
Eurofins DiscoverX DMPK Protein, Active
Active, 6His-tagged, recombinant, human DMPK, amino acids 1-549, for use in Kinase Assays.
Brand: Eurofins DiscoverX 14-649M
Description
Myotonic dystrophy (DM1) is the most common form of muscular dystrophy in adults, caused by amplification of an unstable (CTG)n repeat in the 3' untranslated region (3'-UTR) of the DM1 protein kinase (DMPK) gene. Myotonic dystrophy protein kinase (DMPK) is a serine threonine kinase encoded by the DM-1 locus.
GenBank NM_004409
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
250 μg | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
≥66% |
65 kDa | |
Human | |
N-terminal 6His | |
Truncated | |
DMPK, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.