Learn More
Invitrogen™ eBioscience™ Human Intracellular (nuclear/transcription factor) Protein Flow Cytometry Workflow Kit
The eBioscience™ Human Intracellular (nuclear/transcription factor) Protein Flow Cytometry Workflow Kit contains all of the normal buffers, blockers, and beads needed to complete a standard flow cytometry experiment for staining intracellular nuclear and transcription factor targets in human cell suspensions.
Brand:  Invitrogen™ A53016
Product Code. 17727473
  • 4335.00 SEK / Each
Explore more special offers
Description
Description
  • Kit contains all of the normal buffers, blockers, and beads to complete a standard flow cytometry experiment for staining intracellular nuclear and transcription factor targets in human cell suspensions
  • When combined with experiment-specific antibodies, the reagents within this kit have been optimized to work together to detect human intracellular nuclear and transcription targets by flow cytometry
  • Effective compensation controls
  • Fc receptor blocking
  • Non-antibody, non-protein based blocking solution for reducing background staining from fluorophores
  • Antibody binding buffer which minimizes non-specific binding of antibodies
  • Staining buffer optimized for transcription factors and nuclear proteins
Kit Includes
  • UltraComp eBeads™ Plus Compensation Beads (01-3333-41)
  • Fc Receptor Binding Inhibitor Polyclonal Antibody, eBioscience (14-9161-71)
  • CellBlox™ Blocking Buffer (B001T06F01)
  • eBioscience Foxp3/Transcription Factor Staining Buffer Set (00-5523-00)
  • eBioscience™ Flow Cytometry Staining Buffer (00-4222-57)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Flow Cytometry