Läs mer
Eurofins DiscoverX EGFR (T790M, L858R) Protein, Active
Beskrivning
Receptors that are tyrosine protein kinases include those receptors for epidermal growth factor (EGF), platelet-derived growth factor (PDGF), insulin, and certain lymphokines. Receptor tyrosine kinases are cell surface transmembrane proteins responsible for intracellular signal transduction. They are expressed in many cell types and, after activation by growth factor binding, trigger a series of intracellular pathways leading to a wide variety of responses; e.g. differentiation, proliferation, migration and invasion, angiogenesis, survival. EGF receptor agonism may facilitate wound healing, enhance viral infections, topical inflammation and asthma as well as tumor cell proliferation and metastasis.
GenBank X00588
Specifikationer
Specifikationer
| Formulering | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molekylvikt (g/mol) | 85.8 kDa |
| Kvantitet | 10 μg |
| Art | Human |
| Rekombinant | Recombinant |
| Protein Tag | N-terminal GST |
| Uttryckssystem | Recombinant enzyme expressed in Sf21 insect cells |
| Proteinform | Truncated |
| Renhet eller kvalitetsklass | ≥54% |
| Protein | EGFR (T790M,L858R), active |
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.