missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Epithelial cell marker Antibody (LH39), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2690H
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Epithelial cell marker Monoclonal specifically detects Epithelial cell marker in Primate samples. It is validated for Western Blot.
Specifikationer
| Epithelial cell marker | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| MUC1 | |
| Cells from a single cell suspension of epidermal cells (obtained from fresh human neonatal foreskin) were lysed in Nonidet P40 in phosphate buffered saline and the insoluble pellet was sonicated to prepare insoluble fractions. | |
| 0.1 mL | |
| Cancer, Cellular Markers, Extracellular Matrix | |
| 4582 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| LH39 | |
| Western Blot, Immunohistochemistry | |
| Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Primate | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering