missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol 95% Analytical Grade, Solveco
Click to view available options
Quantity:
12 x 1 L
Packaging:
HDPE Bottle
Description
Denatured Ethanol 95% is a clear, colorless, volatile liquid with a distinctive odor. It reacts explosively with strong oxidizing agents and can react violently with metals to produce hydrogen. It may damage gaskets, painted surfaces, protective and sealing grease coatings, materials, rubber and some plastics.
- Molecular formula: CH3CH2OH
- CAS: 64-17-5
Specifications
Specifications
CAS | 64-17-5 |
Color | Colorless |
Density | 0.8084g/mL at 20°C |
Quantity | 12 x 1 L |
Packaging | HDPE Bottle |
Molecular Formula | C2H6O |
Water | 6.5% max |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
SMILES | CCO |
Show More |
Safety and Handling
Product Identifier
- Etanol 95%
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction