Learn More
Ethyl isobutyrate, 99%, Thermo Scientific Chemicals
507.67 SEK - 1266.21 SEK
Chemical Identifiers
CAS | 97-62-1 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.18 |
MDL Number | MFCD00009165 |
InChI Key | WDAXFOBOLVPGLV-UHFFFAOYSA-N |
Synonym | ethyl isobutyrate, ethyl isobutanoate, ethylisobutyrate, isobutyric acid ethyl ester, ethyl 2,2-dimethylacetate, propanoic acid, 2-methyl-, ethyl ester, isobutyric acid, ethyl ester, ethyl 2-methylpropionate, propionic acid, 2-methyl-, ethyl ester, ethyl isobutyrate natural |
PubChem CID | 7342 |
ChEBI | CHEBI:87303 |
IUPAC Name | ethyl 2-methylpropanoate |
SMILES | CCOC(=O)C(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10472261
|
Thermo Scientific Acros
310515000 |
500 mL | Glass bottle |
507.67 SEK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15169263
|
Thermo Scientific Acros
310510025 |
2.5 L | Glass bottle |
1266.21 SEK
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
97-62-1 | |
116.18 | |
WDAXFOBOLVPGLV-UHFFFAOYSA-N | |
7342 | |
ethyl 2-methylpropanoate |
C6H12O2 | |
MFCD00009165 | |
ethyl isobutyrate, ethyl isobutanoate, ethylisobutyrate, isobutyric acid ethyl ester, ethyl 2,2-dimethylacetate, propanoic acid, 2-methyl-, ethyl ester, isobutyric acid, ethyl ester, ethyl 2-methylpropionate, propionic acid, 2-methyl-, ethyl ester, ethyl isobutyrate natural | |
CHEBI:87303 | |
CCOC(=O)C(C)C |
Specifications
97-62-1 | |
0.8600g/mL | |
13°C | |
99% | |
(CH3)2CHCO2C2H5 | |
MFCD00009165 | |
0.86 | |
Solubility in water: not miscible | |
CCOC(=O)C(C)C | |
116.18 | |
CHEBI:87303 | |
99% | |
Ethyl isobutyrate, 99% |
Colorless | |
112.0°C to 113.0°C | |
Authentic | |
C6H12O2 | |
1.3860 to 1.3880 | |
15,3869 | |
ethyl isobutyrate, ethyl isobutanoate, ethylisobutyrate, isobutyric acid ethyl ester, ethyl 2,2-dimethylacetate, propanoic acid, 2-methyl-, ethyl ester, isobutyric acid, ethyl ester, ethyl 2-methylpropionate, propionic acid, 2-methyl-, ethyl ester, ethyl isobutyrate natural | |
WDAXFOBOLVPGLV-UHFFFAOYSA-N | |
ethyl 2-methylpropanoate | |
7342 | |
116.18 | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-595-4
missing translation for 'rtecsNumber' : NQ4675000
missing translation for 'tsca' : TSCA
RUO – Research Use Only