missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Lactate, 98+%, Extra Pure, SLR, Fisher Chemical

Product Code. 10192040
Click to view available options
Quantity:
2.5 L
Packaging:
Amber glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 97-64-3
Melting Point -23°C
Color Colorless
Boiling Point 154°C
Packaging Amber glass bottle
Molecular Formula C5H10O3
MDL Number 65359
Quantity 2.5 L
Synonym ethyl lactate, actylol, acytol, solactol, lactic acid, ethyl ester, propanoic acid, 2-hydroxy-, ethyl ester, lactate d'ethyle, 2-hydroxypropanoic acid ethyl ester, ethyl 2-hydroxypropionate, ethyl alpha-hydroxypropionate
InChI Key LZCLXQDLBQLTDK-UHFFFAOYSA-N
SMILES CCOC(=O)C(C)O
IUPAC Name ethyl 2-hydroxypropanoate
Molecular Weight (g/mol) 118.132
PubChem CID 7344
ChEBI CHEBI:78321
Formula Weight 118.0682g/mol
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl lactate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : ethyl lactate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'