Läs mer
Thermo Scientific™ F1-ClipTip™ Variabel volym enkanalspipetter
Description
Säkerställ en säker passform med ClipTip teknologi, som låser spetsarna stadigt till pipetten med en fullständig försegling så att de inte lossnar eller faller av. Lättviktshandtagsdesign kombinerat med minimal spetsfästning och utstötningskrafter kommer att öka anpassningen vid pipettering. Känn skillnaden.
Worry-free pipetting is finally here with the only tip that clips.
Sealed in security of ClipTip Technology. Feel the difference.
Eliminate the frustration of banging tips on your pipette
With a light touch, each tip is clipped securely on the pipette.
Reduce the risk of loose or leaking tips
When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles.
Enable consistent and reproducible pipetting from user to user
Higher-quality results and more efficient research. Learn more
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of repetitive strain injuries
Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Improved small volume pipetting
New 0.1-2 μL model and improved versions 1-10 μL models allow optimized performance in your small volume applications.
Wide Selection of ClipTip Pipette Tips
Compatible sizes with F1-ClipTip pipettes: 12.5 Ext, 20, 50, 200, 300, 300 Ext and 1000μL
Life-time Warranty
With convenient online registration, and may vary by country.
Specifications
Specifications
| Kompatibla tips | Klipptips 20 |
| Beskrivning | F1-ClipTip |
| Färg | Pink |
| För användning med | ClipTip™ 20 |
| Antal kanaler | 1 |
| Produktlinje | F1-ClipTip |
| Typ | Variable Volume Pipette |
| Volym (metrisk) | 1 till 10 μl |
| Ökar | 0.02μL |
| Färgkod | Pink |