Läs mer
Cytiva Factor Xa
Produktkod. 10339874 Handla allt Cytiva Produkter
Klicka för att se tillgängliga alternativ
:
400 units
Produktkod. 10339874
Brand: Cytiva 27084901
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Enables the site-specific cleavage of fusion proteins containing an accessible Factor Xa protease recognition sequence such as those expressed in pGEX-X vectors
  • Can be used either following affinity purification or while fusion proteins are bound to Glutathione Sepharose™ media
  • Purified from bovine plasma
  • Specifically cleaves at tetrapeptide Ile-Glu-Gly-Arg
TRUSTED_SUSTAINABILITY
Specifikationer
Produkttyp Facto r Xa Protease
Kvantitet 400U
Kolumntyp Spin Column
För användning med (applikation) Site-specific separation of the GST tag from Proteins expressed using pGEX-X vecto rs, specifically cleaves at Tetrapeptide Ile-Glu-Gly-Arg, one unit cleaves >90% of 100μg of a test GST fusion Protein when incubated in