Läs mer
Falcon™ In Vitro Fertilization Plasticware
Handla allt Corning Produkter
Typ:
17 x 11 mm platta med fyra brunnar
50 x 9 mm lågväggig parabol
60 x 15 mm rund skål
60 x 15 mm skål med en brunn
Kulturområde:
1,39 cm2
19 cm2
2,89 cm2
21,29 cm2
Ytbehandling:
Inte TC-behandlad
TC-behandlad
Kvantitet:
1/förpackning, 100/förpackning
20/förpackning, 500/förpackning
4/paket, 100/förpackning
Förpackningsstorlek:
Paket med 100
Paket med 500
Beskrivning
Note: The lids of the Falcon™ IVF Products are not certified nonembryotoxic.
- Pre-tested plasticware for in vitro fertilization (IVF)
- CE marked to the Medical Device Directive 93/42/EEC
- Manufactured in accordance with FDA Quality System Regulation
- US FDA-cleared Class II medical device
- Nonembryotoxic, Nonpyrogenic, Noncytotoxic
- Tissue culture (TC)-treated for a consistent hydrophilic surface
- Low-wall dish is not TC-treated (not wettable) for optimal media droplet consistency
- Sterilized by gamma irradiation (SAL 10-6)
- Flat dish bottoms for distortion-free optics
- Packaged in peel-open, medical-style packaging, Multi-unit bags have reseal tabs
Certifikat
CE marked
Specifikationer
Specifikationer
| Kulturområde | 19 cm2 |
| Material | Polystyrene |
| Typ | 50 x 9 mm lågväggig parabol |
| Kvantitet | 4/paket, 100/förpackning |
| Antal per förpackning | 4/Pk. |
| nr per ärende | 100/Cs. |
| Ytbehandling | Inte TC-behandlad |
| Sterilitet | Sterile |