Learn More
Novus Biologicals™ FEZ2 Overexpression Lysate
Brand:  Novus Biologicals™ NBP2-07794
Product Code. 18038567
  • 6785.00 SEK / 0.10mg
Explore more special offers
Description
Description
The FEZ2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_593. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens FEZ2, mRNA.
Native
Special Offers
For Research Use Only