Läs mer
Fisherbrand™ Hands-Free Fold-Up Magnifiers
Handla allt Fisherbrand Produkter
Förstoring:
10 gånger
5x
Diameter (engelsk) Lins:
0,5 tum
1,25 tum
Förpackningsstorlek:
Varje
Beskrivning
Innehåller
Batteries, with lighted models.
Fisher Scientific™ Hands-Free Fold-Up Magnifiers all fold twice for compact storage. Allow two-handed examination of objects while yielding a sharp, crisp and distinct image.
- Bottom panel features white grid markings in both inches and millimeters to assist in measuring
- All-glass optics ensure perfect focus with zero distortion
- Seven sizes are available, including three featuring brilliant battery-powered illumination
Specifikationer
Specifikationer
| För användning med (applikation) | For use in quality control, labs, food, electronics, manufacturing and universities |
| Diameter (engelsk) Lins | 1,25 tum |
| Diameter (metrisk) lins | 3.17 cm |
| Förstoring | 5x |
| Material | Glass |
| Produkttyp | Magnifier (Hands-free, Fold-up) |