Läs mer
Fisherbrand™ Accumet™ Temperature-Compensated Two-Cell Conductivity Probes
Handla allt Fisherbrand Produkter
Kontakttyper:
8-stift
Förpackningsstorlek:
Varje
Beskrivning
Fisherbrand™ Accumet™ Temperature-Compensated Two-Cell Conductivity Probes - Mercury - Free, k=10cm-1, 1 to 200 mS/cm, with 8 pin connector
Description
For use on Accumet™ Excel (XL) series benchtop meters
- Immersion-type cell
- Use in standard electrode holder: unique compact size is same as typical pH electrode
- Built-in temperature sensors provide temperature compensated readings
- Epoxy body
Specifikationer
Specifikationer
| Cell konstant | 10 |
| Kontakttyper | 8-stift |
| Sensortyper | Konduktivitet |
| Temperaturnoggrannhet | Temperature Compensated |
| Typ | Konduktivitetssond |
| Material | Epoxy |
| Mätområden | 1 till 200 mS/cm |