Learn More
Proteintech™ FlexAble CoraLite™ Plus 650 Antibody Labeling Kit for Mouse IgG1
This Antibody Labeling Kit conjugates CoraLite™ Plus 650 into mouse IgG1 primary antibodies.
Brand: Proteintech™ KFA023-200
This item is not returnable.
View return policy
Description
FlexAble CoraLite™ Plus 650 Antibody Labeling Kit for Mouse IgG1 is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 650 in any buffer condition to mouse IgG1 primary antibodies from any supplier.
- One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
- Label your mouse IgG1 antibody in <10 min without any additional equipment
Specifications
4 x 50 μL CoraLite™ Plus 650 FlexLinker for Mouse IgG1, 4 x 100 μL FlexQuencher for Mouse IgG1, 4 x 500 μL FlexBuffer, Manual, Protocol Magnet | |
1 Year at -20°C or 6 Months at +4°C, Avoid Light | |
IgG1 |
200 x 0.5 μg Reactions | |
654 nm/674 nm | |
Mouse |