Learn More
MP Biomedicals™ Fluorescent Brightener 28
Fluoresces clear bluish under UV radiation. Fluorescent Brightener 28 is a colorless organic compound that is used for the staining of fungi and as a viability stain. Industrial uses include use as a fluorescent brightening agent for cellulose and polyamide fabrics, paper and detergents and soaps.
848.00 SEK - 11771.30 SEK
Specifications
Storage Requirements | Store at Room Temperature (15°C to 30°C) |
---|---|
Format | Powder |
Type | Fluorescent Brightener |
Concentration | 0.05% to 0.20% |
For Use With (Application) | Fluorescent Brightener 28 is used for the staining of fungi and as a viability stain. Industrial uses include use as a fluorescent brightening agent for cellulose and polyamide fabrics, paper and in detergents and soaps. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11445501
|
MP Biomedicals™
0215806701 |
1 g |
848.00 SEK
1g |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
11455501
|
MP Biomedicals™
0215806705 |
5 g |
2862.00 SEK
5g |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
11465501
|
MP Biomedicals™
0215806725 |
25 g |
11771.30 SEK
25g |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
Description
Fluoresces clear bluish under UV radiation. Fluorescent Brightener 28 is a colorless organic compound that is used for the staining of fungi and as a viability stain. Industrial uses include use as a fluorescent brightening agent for cellulose and polyamide fabrics, paper and detergents and soaps.
- This product is related structurally to Fluorescent Brightner 9 (Calcofluor White MR), which was used by Jefferies and Belcher and by Maeda and Ischida in plant tissue studies.
Specifications
Store at Room Temperature (15°C to 30°C) | |
Fluorescent Brightener | |
Fluorescent Brightener 28 is used for the staining of fungi and as a viability stain. Industrial uses include use as a fluorescent brightening agent for cellulose and polyamide fabrics, paper and in detergents and soaps. | |
290°C | |
C40H42N12O10S2Na2 | |
Calcofluor White M2R, Tinopal UNPA-GX |
Powder | |
0.05% to 0.20% | |
Yellow | |
4404-43-7 | |
960.96 |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.