Läs mer
Gallium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 047148.14
| Kvantitet | 25g |
|---|
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ampouled under argon
Kemiska identifierare
| 13450-91-4 | |
| 450.436 | |
| DWRNSCDYNYYYHT-UHFFFAOYSA-K | |
| 83478 | |
| [Ga](I)(I)I |
| GaI3 | |
| MFCD00016107 | |
| gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basis | |
| triiodogallane |
Specifikationer
| Gallium(III) iodide | |
| 13450-91-4 | |
| (metals basis) | |
| MFCD00016107 | |
| gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basis | |
| DWRNSCDYNYYYHT-UHFFFAOYSA-K | |
| triiodogallane | |
| 83478 | |
| 99.999% | |
| Light and moisture sensitive |
| Ultra dry | |
| 25g | |
| GaI3 | |
| UN3260 | |
| Decomposes in water. | |
| [Ga](I)(I)I | |
| 450.436 | |
| 450.43 | |
| Ampule | |
| 4.15 g/mL |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.