Läs mer
Gilson™ PIPETMAN™ Neo Pipets
Handla allt Gilson Produkter
Noggrannhet:
±0.024, ±0.025, ±0.03μL
±0.1, ±0.1, ±0.2μL
±0.35, ±0.4, ±0.8μL
±0.5, ±0.8, ±1.6μL
±3, ±4, ±8μL
Modell:
P1000N
P100N
P200N
P20N
P2N
Volym (metrisk):
0,2 till 2 μl
10 till 100 μl
100 till 1000 μl
2 till 20 μl
20 till 200 μl
Kompatibla tips:
D10, DL10, DF10ST, DFL10ST
D1000, DF1000ST, DF1200ST
D200, D300, DF100ST, DF200ST, DF300ST
D200, DF100ST
D200, DF30ST
Förpackningsstorlek:
Varje
Beskrivning
- Redesigned tip ejector and ejection springs reduces tip ejection forces
- Unique clip ejector fits securely into the body of the pipette to avoid slipping over time
- PVDF handle limits the transfer of heat from hand to the pipette and is resistant to aggressive chemicals
- Stainless steel tip ejector resists corrosion and offers a smooth tip ejection stroke
- Color-coded push-button for ease of identification
- Unique tip holder allows for a secure fit of most brands of tip
- Models P2N and P10N incorporate dual position tip ejectors, allowing the use of D10 (short) and DL10 (long)
Beställningsinfo
Trade in program available, please contact your Fisher Scientific Sales representative for more details.
Specifikationer
Specifikationer
| Noggrannhet | ±0.1, ±0.1, ±0.2μL |
| Certifikat och efterlevnad | ISO8655 |
| Kompatibla tips | D200, DF30ST |
| Modell | P20N |
| Antal kanaler | 1 |
| Produktlinje | Pipetman Neo |
| Typ | Pipette |
| Volym (metrisk) | 2 till 20 μl |
| Finger stil | Ergonomic |
| Inkluderar | Color coded push button, PVDF handle, Stainless Steel tip ejectors, universal tip holder |