Learn More
Jaytec Glass™ Glass Capillary Tubes
These Glass Capillary Tubes use a liquid's properties of cohesion, adhesion and surface tension to draw liquid up the interior walls of a narrow tube.
Brand:  Jaytec Glass™ CAPSPECMERC
 View more versions of this product
Product Code. 12326297
  • 1600.00 SEK / Pack of 1000
Explore more special offers
Description
Description
  • Manufactured from industry-standard soda glass
Specifications
Specifications
Soda Glass
Capillary Tubes
75 mm
1 μL
0.48 to 0.5 mm
1000 Pk.