Learn More
Gilac™ HACCP Cook Chill Crate
HACCP Cook Chill Crate is a 600 x 400 perforated crate, ideal for commercial kitchen use and the cold chain process. It provides ventilation and is stacking and nesting when half turned.
Brand: Gilac™ G180321-1
This item is not returnable.
View return policy
Description
- 600 x 400 perforated crate.
- Ideal for commercial kitchen use and the cold chain process.
- Provides ventilation.
- Stacking and nesting when half turned.
- Large perforations, meaning that temperatures can be brought down quickly and it is easy to clean.
- Crates available in 5 HACCP colors.
- Tracking management thanks to a contents identification label (IML): In mold labelling process whereby the label is injection-molded into the plastic rendering it permanent and rewritable.
- Use with a water erasable marking pen.
Specifications
9.8 gal. | |
None | |
HDPE | |
Crate | |
600 mm | |
400 mm | |
197 mm |
37 L | |
White | |
Rectangular | |
600 x 400 x 197 mm | |
23.6 in. | |
15.7 in. | |
7.8 in. |