Learn More
Gilac™ HACCP Cook Chill Crate
HACCP Cook Chill Crate is a 600 x 400 perforated crate, ideal for commercial kitchen use and the cold chain process. It provides ventilation and is stacking and nesting when half turned.
Brand:  Gilac™ G180310-1
 View more versions of this product
Product Code. 17928511
  • 383.00 SEK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • 600 x 400 perforated crate.
  • Ideal for commercial kitchen use and the cold chain process.
  • Provides ventilation.
  • Stacking and nesting when half turned.
  • Large perforations, meaning that temperatures can be brought down quickly and it is easy to clean.
  • Crates available in 5 HACCP colors.
  • Tracking management thanks to a contents identification label (IML): In mold labelling process whereby the label is injection-molded into the plastic rendering it permanent and rewritable.
    • Use with a water erasable marking pen.
Specifications
Specifications
9.8 gal.
None
HDPE
Crate
600 mm
400 mm
197 mm
37 L
Blue
Rectangular
600 x 400 x 197 mm
23.6 in.
15.7 in.
7.8 in.