Learn More
Gilac™ HACCP Cook Chill Crate
HACCP Cook Chill Crate is a 600 x 400 perforated crate, ideal for commercial kitchen use and the cold chain process. It provides ventilation and is stacking and nesting when half turned.
366.00 SEK - 383.00 SEK
Specifications
Closure Type | None |
---|---|
Material | HDPE |
Shape | Rectangular |
Product Type | Crate |
Length (Metric) | 600 mm |
Product Code | Brand | Capacity (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Color | Price | Quantity & Availability | ||||
17948641
|
Gilac™
G180210-1 |
27 L | Blue |
366.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17998821
|
Gilac™
G180211-1 |
27 L | Green |
366.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17988421
|
Gilac™
G180218-1 |
27 L | Yellow |
366.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17958571
|
Gilac™
G180221-1 |
27 L | White |
366.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17958641
|
Gilac™
G180209-1 |
27 L | Red |
366.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17998421
|
Gilac™
G180309-1 |
37 L | Red |
383.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17928511
|
Gilac™
G180310-1 |
37 L | Blue |
383.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17988821
|
Gilac™
G180311-1 |
37 L | Green |
383.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17908831
|
Gilac™
G180318-1 |
37 L | Yellow |
383.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17918831
|
Gilac™
G180321-1 |
37 L | White |
383.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- 600 x 400 perforated crate.
- Ideal for commercial kitchen use and the cold chain process.
- Provides ventilation.
- Stacking and nesting when half turned.
- Large perforations, meaning that temperatures can be brought down quickly and it is easy to clean.
- Crates available in 5 HACCP colors.
- Tracking management thanks to a contents identification label (IML): In mold labelling process whereby the label is injection-molded into the plastic rendering it permanent and rewritable.
- Use with a water erasable marking pen.
Specifications
None | |
Rectangular | |
600 mm | |
400 mm |
HDPE | |
Crate | |
23.6 in. | |
15.7 in. |