Learn More
Gilac™ HACCP Cook Chill Crate
HACCP Cook Chill Crate is a 600 x 400 perforated crate, ideal for commercial kitchen use and the cold chain process. It provides ventilation and is stacking and nesting when half turned.
366.00 SEK - 383.00 SEK
Specifications
Closure Type None
Material HDPE
Shape Rectangular
Product Type Crate
Length (Metric) 600 mm
View More Specs
Products 10
Product Code Brand Capacity (Metric) Color Price Quantity & Availability  
Product Code Brand Capacity (Metric) Color Price Quantity & Availability  
17948641
View Documents
Gilac™
G180210-1
27 L Blue
366.00 SEK
Each
17998821
View Documents
Gilac™
G180211-1
27 L Green
366.00 SEK
Each
17988421
View Documents
Gilac™
G180218-1
27 L Yellow
366.00 SEK
Each
17958571
View Documents
Gilac™
G180221-1
27 L White
366.00 SEK
Each
17958641
View Documents
Gilac™
G180209-1
27 L Red
366.00 SEK
Each
17998421
View Documents
Gilac™
G180309-1
37 L Red
383.00 SEK
Each
17928511
View Documents
Gilac™
G180310-1
37 L Blue
383.00 SEK
Each
17988821
View Documents
Gilac™
G180311-1
37 L Green
383.00 SEK
Each
17908831
View Documents
Gilac™
G180318-1
37 L Yellow
383.00 SEK
Each
17918831
View Documents
Gilac™
G180321-1
37 L White
383.00 SEK
Each
Description
Description
  • 600 x 400 perforated crate.
  • Ideal for commercial kitchen use and the cold chain process.
  • Provides ventilation.
  • Stacking and nesting when half turned.
  • Large perforations, meaning that temperatures can be brought down quickly and it is easy to clean.
  • Crates available in 5 HACCP colors.
  • Tracking management thanks to a contents identification label (IML): In mold labelling process whereby the label is injection-molded into the plastic rendering it permanent and rewritable.
    • Use with a water erasable marking pen.
Specifications
Specifications
None
Rectangular
600 mm
400 mm
HDPE
Crate
23.6 in.
15.7 in.