Learn More
GM MERC Biotech Heat Transfer Fluid for Osmometers and Cryoscopes
Heat transfer fluid (cooling fluid) specially designed for osmometers and cryoscopes. Flask contains 160 mL environmentally safe medical grade cooling liquid: H2O + Propylene Glycole. Heat transfer fluid serves as an intermediary in cooling, transporting and storing thermal energy, and heating.
Brand: GM MERC Biotech 101910
This item is not returnable.
View return policy
Description
- Suitable for all osmometers and cryoscopes
- 160 mL flask (fits into Osmometer Model 3250)
- Manufactured in Denmark
- Medical grade cooling fluid
- Consistently quality controlled on the 3900 and 3250 osmometers
Specifications
Osmometers and Cryoscopes | |
Single-use | |
160 mL |