missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ HEPA Air Filter

For Use With: FD 56

Brand:  BINDER™ 8012-1050

Additional Details : Weight : 0.50000kg

Product Code. 16419123

  • 19194.36 SEK / Each
Estimated Shipment: 15-10-2024
to see stock.
Add to basket

Description

Description

  • On fresh air supply for unit
  • Filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% at 0.3 μm)
Specifications

Specifications

HEPA air filter - On fresh air supply for unit; filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% (at 0.3 μm))
BINDER Chambers FD 56
HEPA Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers