missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexan-1-ol, Extra Pure, SLR, Fisher Chemical™
655.00 SEK - 5685.00 SEK
Chemical Identifiers
CAS | 111-27-3 |
---|---|
Molecular Formula | C6H14O |
Molecular Weight (g/mol) | 102.177 |
MDL Number | 2982 |
InChI Key | ZSIAUFGUXNUGDI-UHFFFAOYSA-N |
Synonym | 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol |
PubChem CID | 8103 |
ChEBI | CHEBI:87393 |
IUPAC Name | hexan-1-ol |
SMILES | CCCCCCO |
Description
Chemical Identifiers
111-27-3 | |
102.177 | |
ZSIAUFGUXNUGDI-UHFFFAOYSA-N | |
8103 | |
hexan-1-ol |
C6H14O | |
2982 | |
1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol | |
CHEBI:87393 | |
CCCCCCO |
Specifications
111-27-3 | |
Colorless | |
C6H14O | |
1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol | |
CCCCCCO | |
102.177 | |
CHEBI:87393 | |
Liquid |
-52°C | |
156°C | |
2982 | |
ZSIAUFGUXNUGDI-UHFFFAOYSA-N | |
hexan-1-ol | |
8103 | |
102.18g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Hexyl alcohol
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.