Learn More
Thermo Scientific Chemicals High Form Nickel Crucible
372.76 SEK - 2863.77 SEK
Specifications
Description | High Form |
---|---|
Material | Nickel |
Product Type Specs | Nickel Crucible |
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | |||
15493575
|
Thermo Scientific Chemicals
035883.KT |
20 mL | 38mm | 35mm |
372.76 SEK
Each |
In Stock
Log in to see stock. |
|||
15463585
|
Thermo Scientific Chemicals
035904.KT |
30 mL | 41mm | 43mm |
648.49 SEK
Each |
In Stock
Log in to see stock. |
|||
15423595
|
Thermo Scientific Chemicals
035925.KT |
50 mL | 44mm | 51mm |
743.16 SEK
Each |
In Stock
Log in to see stock. |
|||
15483595
|
Thermo Scientific Chemicals
035946.KT |
75 mL | 54mm | 57mm |
657.96 SEK
Each |
In Stock
Log in to see stock. |
|||
15443605
|
Thermo Scientific Chemicals
035967.KT |
100 mL | 61mm | 62mm |
1020.07 SEK
Each |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||
15403625
|
Thermo Scientific Chemicals
036019.KT |
250 mL | 84mm | 78mm |
1372.72 SEK
Each |
In Stock
Log in to see stock. |
|||
15483625
|
Thermo Scientific Chemicals
036040.KT |
500 mL | 101mm | 91mm |
2863.77 SEK
Each |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
High Form | |
Nickel | |
Nickel Crucible |
RUO – Research Use Only